Learn More
Meso Scale Discovery UPLEX HUM CD276/B7H3 ASYQP
SDP
Catalog No. 5000080189
Encompass
The Meso Scale Discovery U-PLEX Human CD276 (B7-H3) Assay is a multiplex immunoassay kit for quantifying human CD276 protein in various samples This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection of CD276 levels down to pg/mL The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators controls diluent read buffer and necessary reagents It enables high-throughput reproducible analysis of CD276 a key immune checkpoint molecule
Catalog No. 50-000-80189 Supplier Meso Scale Discovery Supplier No. K151E7K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human CD276 (B7-H3) Assay is a multiplex immunoassay kit for quantifying human CD276 protein in various samples This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection of CD276 levels down to pg/mL The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators controls diluent read buffer and necessary reagents It enables high-throughput reproducible analysis of CD276 a key immune checkpoint molecule