Learn More
Meso Scale Discovery UPLEX HUM CD276/B7HASY384
SDP
Catalog No. 5000081007
Encompass
The Meso Scale Discovery CD276 (B7-H3) Kit is a sensitive electrochemiluminescent immunoassay for quantifying CD276 (B7-H3) in human serum plasma and cell culture supernatants It uses SULFO-TAG labels that emit light when stimulated via carbon electrodes The kit includes a 96-well plate with a CD276 capture antibody a detection antibody with a SULFO-TAG label and calibrators and controls for a standard curve This format allows accurate CD276 quantification in small samples providing a rapid reproducible method for research use
Catalog No. 50-000-81007 Supplier Meso Scale Discovery Supplier No. K251E7K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery CD276 (B7-H3) Kit is a sensitive electrochemiluminescent immunoassay for quantifying CD276 (B7-H3) in human serum plasma and cell culture supernatants It uses SULFO-TAG labels that emit light when stimulated via carbon electrodes The kit includes a 96-well plate with a CD276 capture antibody a detection antibody with a SULFO-TAG label and calibrators and controls for a standard curve This format allows accurate CD276 quantification in small samples providing a rapid reproducible method for research use