Learn More
Meso Scale Discovery UPLEX HUM CD28 ASY384well
SDP
Catalog No. 5000080699
Encompass
The Meso Scale Discovery U-PLEX Human CD28 384-Well Assay Kit is a multiplex immunoassay that measures CD28 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay allowing high throughput analysis of up to 80 samples The assay has a detection range of 50-10 000 pg/mL and includes reagents for a 384-well plate including antibodies calibrators controls assay diluent read buffer and wash buffer This quantitative immunoassay allows sensitive and accurate measurement of CD28 which plays a role in T cell activation
Catalog No. 50-000-80699 Supplier Meso Scale Discovery Supplier No. K251H7K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human CD28 384-Well Assay Kit is a multiplex immunoassay that measures CD28 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay allowing high throughput analysis of up to 80 samples The assay has a detection range of 50-10 000 pg/mL and includes reagents for a 384-well plate including antibodies calibrators controls assay diluent read buffer and wash buffer This quantitative immunoassay allows sensitive and accurate measurement of CD28 which plays a role in T cell activation