Learn More
Meso Scale Discovery UPLEX HUM CD28 ASYQP 25 PL
SDP
Catalog No. 5000082479
Encompass
The U-PLEX Human CD28 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures CD28 levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrator diluent wash buffer read buffer and a vial of lyophilized calibrator The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-82479 Supplier Meso Scale Discovery Supplier No. K151H7K24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human CD28 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures CD28 levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrator diluent wash buffer read buffer and a vial of lyophilized calibrator The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL