Learn More
Meso Scale Discovery UPLEX HUM CD40L soluble ASY
SDP
Catalog No. 5000080800
Encompass
The Meso Scale Discovery U-PLEX Human CD40L (soluble) Assay Kit is designed for the quantitative measurement of human CD40 ligand in serum plasma cell culture supernatants cell and tissue lysates This electrochemiluminescent multiplex assay kit allows for the simultaneous measurement of CD40L and other biomarkers It utilizes MSD s proprietary SULFO-TAG labels and provides high sensitivity broad dynamic range low sample consumption and rapid time to results This assay requires an MSD instrument such as QuickPlex SQ 120 or HD-1 for analysis The kit includes a 96-well plate coated with capture antibodies detection antibodies and calibrators
Catalog No. 50-000-80800 Supplier Meso Scale Discovery Supplier No. K151L7K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human CD40L (soluble) Assay Kit is designed for the quantitative measurement of human CD40 ligand in serum plasma cell culture supernatants cell and tissue lysates This electrochemiluminescent multiplex assay kit allows for the simultaneous measurement of CD40L and other biomarkers It utilizes MSD s proprietary SULFO-TAG labels and provides high sensitivity broad dynamic range low sample consumption and rapid time to results This assay requires an MSD instrument such as QuickPlex SQ 120 or HD-1 for analysis The kit includes a 96-well plate coated with capture antibodies detection antibodies and calibrators