Learn More
Meso Scale Discovery UPLEX HUM CD40L soluble ASY
SDP
Catalog No. 5000079550
Encompass
The Meso Scale Discovery U-PLEX Human CD40L (soluble) Assay Kit is a multiplex immunoassay that measures soluble CD40 ligand in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent The assay has a typical lower limit of detection of 0 08 ng/mL and an upper limit of quantification of 2500 pg/mL
Catalog No. 50-000-79550 Supplier Meso Scale Discovery Supplier No. K151L7K1
The Meso Scale Discovery U-PLEX Human CD40L (soluble) Assay Kit is a multiplex immunoassay that measures soluble CD40 ligand in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent The assay has a typical lower limit of detection of 0 08 ng/mL and an upper limit of quantification of 2500 pg/mL