Learn More
Meso Scale Discovery UPLEX HUM CD40L soluble ASY384
SDP
Catalog No. 5000079571
Encompass
The Meso Scale Discovery U-PLEX Human CD40L (soluble) 384-well assay is a multiplex immunoassay panel that measures soluble CD40 ligand in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology in a 384-well plate format providing high sensitivity and a broad dynamic range This assay allows for the quantitative measurement of soluble CD40L an important biomarker involved in inflammatory and autoimmune diseases With a sample volume of 25 L the assay offers a streamlined and efficient workflow Overall this is a high-performance assay well-suited for biomarker research and drug discovery applications involving soluble CD40L
Catalog No. 50-000-79571 Supplier Meso Scale Discovery Supplier No. K251L7K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human CD40L (soluble) 384-well assay is a multiplex immunoassay panel that measures soluble CD40 ligand in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology in a 384-well plate format providing high sensitivity and a broad dynamic range This assay allows for the quantitative measurement of soluble CD40L an important biomarker involved in inflammatory and autoimmune diseases With a sample volume of 25 L the assay offers a streamlined and efficient workflow Overall this is a high-performance assay well-suited for biomarker research and drug discovery applications involving soluble CD40L