Learn More
Meso Scale Discovery UPLEX HUM CD40L soluble ASYQP
SDP
Catalog No. 5000082348
Encompass
The Meso Scale Discovery U-PLEX Human CD40L (soluble) Assay is a multiplex immunoassay kit that enables simultaneous quantification of CD40L and other biomarkers in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology for high sensitivity and broad dynamic range The assay requires 50 L or less sample volume and can be read on MSD instrument platforms It includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and measure CD40L concentration This assay provides a rapid reproducible method to measure soluble CD40L levels in research studies
Catalog No. 50-000-82348 Supplier Meso Scale Discovery Supplier No. K151L7K21
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX Human CD40L (soluble) Assay is a multiplex immunoassay kit that enables simultaneous quantification of CD40L and other biomarkers in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology for high sensitivity and broad dynamic range The assay requires 50 L or less sample volume and can be read on MSD instrument platforms It includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and measure CD40L concentration This assay provides a rapid reproducible method to measure soluble CD40L levels in research studies