Learn More
Meso Scale Discovery UPLEX HUM CD40L soluble ASYQP
SDP
Catalog No. 5000079551
Encompass
The Meso Scale Discovery U-PLEX Human CD40L (soluble) Assay is an electrochemiluminescent immunoassay for the quantitative measurement of soluble CD40 ligand in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible results The assay has a lower limit of detection of 0 05 ng/mL and an upper limit of quantification of 10 000 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody labeled with MSD SULFO-TAG calibrators and sample diluent The total assay time is 3 5 hours
Catalog No. 50-000-79551 Supplier Meso Scale Discovery Supplier No. K151L7K22
The Meso Scale Discovery U-PLEX Human CD40L (soluble) Assay is an electrochemiluminescent immunoassay for the quantitative measurement of soluble CD40 ligand in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible results The assay has a lower limit of detection of 0 05 ng/mL and an upper limit of quantification of 10 000 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody labeled with MSD SULFO-TAG calibrators and sample diluent The total assay time is 3 5 hours