Learn More
Meso Scale Discovery UPLEX HUM CD40L soluble ASYQP
SDP
Catalog No. 5000082349
Encompass
The Meso Scale Discovery U-PLEX Human CD40L (soluble) Assay is an electrochemiluminescent multiplex immunoassay designed to measure soluble CD40 ligand in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to enable the simultaneous detection of multiple protein biomarkers in a single well This assay provides high sensitivity up to 5 logs of dynamic range and requires low sample volumes The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and an MSD read buffer It can accurately quantify soluble CD40L levels between 0 05-500 ng/mL
Catalog No. 50-000-82349 Supplier Meso Scale Discovery Supplier No. K151L7K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human CD40L (soluble) Assay is an electrochemiluminescent multiplex immunoassay designed to measure soluble CD40 ligand in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to enable the simultaneous detection of multiple protein biomarkers in a single well This assay provides high sensitivity up to 5 logs of dynamic range and requires low sample volumes The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and an MSD read buffer It can accurately quantify soluble CD40L levels between 0 05-500 ng/mL