Learn More
Meso Scale Discovery UPLEX HUM Clusterin ASY
SDP
Catalog No. 5000078740
Encompass
The Meso Scale Discovery U-PLEX Human Clusterin Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Clusterin in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and a wide dynamic range The assay has high specificity and no cross-reactivity It requires only 50 L sample volume and can be multiplexed with other U-PLEX assays for efficient biomarker analysis The kit provides reagents for 96 tests in a quick 3 5 hour protocol
Catalog No. 50-000-78740 Supplier Meso Scale Discovery Supplier No. K151B9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Clusterin Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Clusterin in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and a wide dynamic range The assay has high specificity and no cross-reactivity It requires only 50 L sample volume and can be multiplexed with other U-PLEX assays for efficient biomarker analysis The kit provides reagents for 96 tests in a quick 3 5 hour protocol