Learn More
Meso Scale Discovery UPLEX HUM Clusterin ASY384
SDP
Catalog No. 5000080277
Encompass
The Meso Scale Discovery U-PLEX Human Clusterin 384-Well Assay is a multiplex immunoassay that measures Clusterin levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This 384-well assay allows high throughput analysis of Clusterin across multiple samples It includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags and calibrator and control samples to generate a standard curve and validate assay performance
Catalog No. 50-000-80277 Supplier Meso Scale Discovery Supplier No. K251B9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Clusterin 384-Well Assay is a multiplex immunoassay that measures Clusterin levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This 384-well assay allows high throughput analysis of Clusterin across multiple samples It includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags and calibrator and control samples to generate a standard curve and validate assay performance