Learn More
Meso Scale Discovery UPLEX HUM Clusterin ASY384well
SDP
Catalog No. 5000081003
Encompass
The Meso Scale Discovery U-PLEX Human Clusterin 384-Well Assay is an electrochemiluminescent immunoassay designed for the quantitative measurement of Clusterin in human serum plasma and cell culture supernatants It utilizes MSD s proprietary U-PLEX technology is provided in a 384-well plate format and has a sample volume of 25 L The assay has a typical sensitivity of 0 3 ng/mL and an upper limit of quantification of 10 000 ng/mL This product can aid in the research of Alzheimer s disease and other neurodegenerative disorders in which Clusterin is a biomarker
Catalog No. 50-000-81003 Supplier Meso Scale Discovery Supplier No. K251B9K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Clusterin 384-Well Assay is an electrochemiluminescent immunoassay designed for the quantitative measurement of Clusterin in human serum plasma and cell culture supernatants It utilizes MSD s proprietary U-PLEX technology is provided in a 384-well plate format and has a sample volume of 25 L The assay has a typical sensitivity of 0 3 ng/mL and an upper limit of quantification of 10 000 ng/mL This product can aid in the research of Alzheimer s disease and other neurodegenerative disorders in which Clusterin is a biomarker