Learn More
Meso Scale Discovery UPLEX HUM Clusterin ASYQP
SDP
Catalog No. 5000078739
Encompass
The Meso Scale Discovery U-PLEX Human Clusterin Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Clusterin in human serum plasma and cell culture supernatants It utilizes MSD s proprietary U-PLEX technology is provided in a convenient all-in-one kit has a detection range of 50-10 000 pg/mL and exhibits high sensitivity specificity and reproducibility The assay requires an MSD instrument such as QuickPlex QuickPlex SQ 120 or V-PLEX to read the electrochemiluminescent signal Overall the U-PLEX Human Clusterin Assay provides a rapid sensitive and reliable tool for quantifying Clusterin levels in biological samples
Catalog No. 50-000-78739 Supplier Meso Scale Discovery Supplier No. K151B9K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Clusterin Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Clusterin in human serum plasma and cell culture supernatants It utilizes MSD s proprietary U-PLEX technology is provided in a convenient all-in-one kit has a detection range of 50-10 000 pg/mL and exhibits high sensitivity specificity and reproducibility The assay requires an MSD instrument such as QuickPlex QuickPlex SQ 120 or V-PLEX to read the electrochemiluminescent signal Overall the U-PLEX Human Clusterin Assay provides a rapid sensitive and reliable tool for quantifying Clusterin levels in biological samples