Learn More
Meso Scale Discovery UPLEX HUM Comp factor D ASY
SDP
Catalog No. 5000079156
Encompass
The Meso Scale Discovery U-PLEX Human Complement Factor D Assay is a multiplex immunoassay kit for quantifying Complement Factor D in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive accurate measurement with high reproducibility The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals requiring only 50 L per well for high-throughput analysis
Catalog No. 50-000-79156 Supplier Meso Scale Discovery Supplier No. K151K9K2
The Meso Scale Discovery U-PLEX Human Complement Factor D Assay is a multiplex immunoassay kit for quantifying Complement Factor D in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive accurate measurement with high reproducibility The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals requiring only 50 L per well for high-throughput analysis