Learn More
Meso Scale Discovery UPLEX HUM Comp factor D ASY
SDP
Catalog No. 5000079548
Encompass
The Meso Scale Discovery U-PLEX Human Complement Factor D Assay is a multiplex immunoassay kit that enables simultaneous quantification of Complement Factor D protein in human serum plasma and cell culture supernatant samples The assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay allows for the accurate measurement of Complement Factor D levels which can aid in the investigation of complement system activation With minimal hands-on time required this kit serves as a simple and rapid tool for complement-related research
Catalog No. 50-000-79548 Supplier Meso Scale Discovery Supplier No. K151K9K1
The Meso Scale Discovery U-PLEX Human Complement Factor D Assay is a multiplex immunoassay kit that enables simultaneous quantification of Complement Factor D protein in human serum plasma and cell culture supernatant samples The assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay allows for the accurate measurement of Complement Factor D levels which can aid in the investigation of complement system activation With minimal hands-on time required this kit serves as a simple and rapid tool for complement-related research