Learn More
Meso Scale Discovery UPLEX HUM Comp factor D ASY384
SDP
Catalog No. 5000079860
Encompass
The Meso Scale Discovery Complement Factor D Kit is an immunoassay that quantitatively measures complement factor D in human serum plasma and other biological samples The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a broad dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and diluent The assay can detect factor D levels between 0 3-10 000 ng/mL and has high specificity with minimal cross-reactivity to related proteins The kit offers a rapid and robust method to measure complement activation via the alternative pathway
Catalog No. 50-000-79860 Supplier Meso Scale Discovery Supplier No. K251K9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Complement Factor D Kit is an immunoassay that quantitatively measures complement factor D in human serum plasma and other biological samples The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a broad dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and diluent The assay can detect factor D levels between 0 3-10 000 ng/mL and has high specificity with minimal cross-reactivity to related proteins The kit offers a rapid and robust method to measure complement activation via the alternative pathway