Learn More
Meso Scale Discovery UPLEX HUM Comp factor D ASYQP
SDP
Catalog No. 5000082344
Encompass
The Meso Scale Discovery U-PLEX Human Complement Factor D Assay is a multiplex immunoassay kit that enables simultaneous quantification of Complement Factor D in human serum plasma urine saliva CSF and cell culture samples It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range low sample consumption and rapid time to results This assay requires an MSD instrument such as QuickPlex QuickPlex SQ 120 or V-PLEX to run the plate and analyze the data It includes a 96-well plate coated with capture antibodies detection antibodies calibrators and controls needed to perform the assay
Catalog No. 50-000-82344 Supplier Meso Scale Discovery Supplier No. K151K9K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Complement Factor D Assay is a multiplex immunoassay kit that enables simultaneous quantification of Complement Factor D in human serum plasma urine saliva CSF and cell culture samples It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range low sample consumption and rapid time to results This assay requires an MSD instrument such as QuickPlex QuickPlex SQ 120 or V-PLEX to run the plate and analyze the data It includes a 96-well plate coated with capture antibodies detection antibodies calibrators and controls needed to perform the assay