Learn More
Meso Scale Discovery UPLEX HUM Comp factor D ASYQP
SDP
Catalog No. 5000079549
Encompass
The Meso Scale Discovery U-PLEX Human Complement Factor D Assay is a multiplex immunoassay kit that measures Complement Factor D levels in human serum plasma and cell culture supernatant samples The assay utilizes electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay allows for the simultaneous measurement of Complement Factor D along with other related biomarkers It can be used for basic research disease-related research and drug discovery and development
Catalog No. 50-000-79549 Supplier Meso Scale Discovery Supplier No. K151K9K22
The Meso Scale Discovery U-PLEX Human Complement Factor D Assay is a multiplex immunoassay kit that measures Complement Factor D levels in human serum plasma and cell culture supernatant samples The assay utilizes electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay allows for the simultaneous measurement of Complement Factor D along with other related biomarkers It can be used for basic research disease-related research and drug discovery and development