Learn More
Meso Scale Discovery UPLEX HUM Complement C9 ASY384
SDP
Catalog No. 5000079864
Encompass
The Meso Scale Discovery Complement Component C9 Kit is an electrochemiluminescent immunoassay designed for the quantitative measurement of Complement Component C9 in human serum plasma and other biological fluids This assay employs an MSD GOLD SULFO-TAG label and is provided in a convenient 96-well plate format The kit includes all necessary reagents and diluents needed to perform testing This product enables sensitive accurate and rapid quantification of Complement Component C9 levels With a broad dynamic range and high-sensitivity it is well-suited for research and drug development applications related to immunology and inflammation
Catalog No. 50-000-79864 Supplier Meso Scale Discovery Supplier No. K251U9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Complement Component C9 Kit is an electrochemiluminescent immunoassay designed for the quantitative measurement of Complement Component C9 in human serum plasma and other biological fluids This assay employs an MSD GOLD SULFO-TAG label and is provided in a convenient 96-well plate format The kit includes all necessary reagents and diluents needed to perform testing This product enables sensitive accurate and rapid quantification of Complement Component C9 levels With a broad dynamic range and high-sensitivity it is well-suited for research and drug development applications related to immunology and inflammation