Learn More
Meso Scale Discovery UPLEX HUM Complement C9 ASY384
SDP
Catalog No. 5000080731
Encompass
The Meso Scale Discovery U-PLEX Human Complement C9 384-Well Assay Kit quantifies Complement C9 in human serum plasma urine cell culture supernatants and other fluids This 384-well electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels offering high sensitivity broad dynamic range and low sample volume needs The kit includes a 384-well plate with capture antibody detection antibody with ruthenium read buffer wash buffer blocker solutions calibrators and controls The assay s quantification range is 0 122 ng/mL to 1000 ng/mL
Catalog No. 50-000-80731 Supplier Meso Scale Discovery Supplier No. K251U9K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Complement C9 384-Well Assay Kit quantifies Complement C9 in human serum plasma urine cell culture supernatants and other fluids This 384-well electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels offering high sensitivity broad dynamic range and low sample volume needs The kit includes a 384-well plate with capture antibody detection antibody with ruthenium read buffer wash buffer blocker solutions calibrators and controls The assay s quantification range is 0 122 ng/mL to 1000 ng/mL