Learn More
Meso Scale Discovery UPLEX HUM Complement C9 ASYQP
SDP
Catalog No. 5000080507
Encompass
The Meso Scale Discovery U-PLEX Human Complement C9 Assay Kit quantifies Complement C9 in human serum plasma urine cell culture supernatants and other fluids This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection down to pg/mL It includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators controls diluents buffers and seals Compatible with MSD s SECTOR Imager and analyzed via Discovery Workbench software it enables high-throughput analysis of Complement C9 in various samples
Catalog No. 50-000-80507 Supplier Meso Scale Discovery Supplier No. K151U9K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Complement C9 Assay Kit quantifies Complement C9 in human serum plasma urine cell culture supernatants and other fluids This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection down to pg/mL It includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators controls diluents buffers and seals Compatible with MSD s SECTOR Imager and analyzed via Discovery Workbench software it enables high-throughput analysis of Complement C9 in various samples