Learn More
Meso Scale Discovery UPLEX HUM Cpeptide ASY384well
SDP
Catalog No. 5000078491
Encompass
The Meso Scale Discovery C-Peptide Kit is an electrochemiluminescent immunoassay for quantifying C-peptide in human serum plasma and other samples It uses MSD s SULFO-TAG labels and multi-array plate technology for ultra-sensitive detection The assay has a dynamic range of 1 5-5000 pg/mL and requires 25 L of sample It shows high specificity minimal cross-reactivity and correlates well with ELISA assays The kit includes a 96-well plate detection antibody calibrators and diluent ideal for diabetes research and drug development
Catalog No. 50-000-78491 Supplier Meso Scale Discovery Supplier No. K2516JK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery C-Peptide Kit is an electrochemiluminescent immunoassay for quantifying C-peptide in human serum plasma and other samples It uses MSD s SULFO-TAG labels and multi-array plate technology for ultra-sensitive detection The assay has a dynamic range of 1 5-5000 pg/mL and requires 25 L of sample It shows high specificity minimal cross-reactivity and correlates well with ELISA assays The kit includes a 96-well plate detection antibody calibrators and diluent ideal for diabetes research and drug development