Learn More
Meso Scale Discovery UPLEX HUM Cpeptide ASY384well
SDP
Catalog No. 5000078629
Encompass
The Meso Scale Discovery U-PLEX Human C-Peptide Assay Kit quantifies C-peptide in human serum plasma and other fluids This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection on the MESO QuickPlex SQ 120 It has a detection limit of 26 pg/mL and quantification up to 20 000 pg/mL The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents C-peptide measurement aids in diagnosing and monitoring diabetes and insulinoma
Catalog No. 50-000-78629 Supplier Meso Scale Discovery Supplier No. K2516JK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human C-Peptide Assay Kit quantifies C-peptide in human serum plasma and other fluids This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection on the MESO QuickPlex SQ 120 It has a detection limit of 26 pg/mL and quantification up to 20 000 pg/mL The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents C-peptide measurement aids in diagnosing and monitoring diabetes and insulinoma