Learn More
Meso Scale Discovery UPLEX HUM Cpeptide ASYQP
SDP
Catalog No. 5000081709
Encompass
The U-PLEX Human C-Peptide Assay (K1516JK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of human C-peptide in biological samples This electrochemiluminescence-based assay offers high sensitivity and specificity for diabetes research and clinical applications The kit provides reliable detection of C-peptide a biomarker for endogenous insulin production enabling researchers to assess pancreatic beta-cell function in various sample types including serum and plasma
Catalog No. 50-000-81709 Supplier Meso Scale Discovery Supplier No. K1516JK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human C-Peptide Assay (K1516JK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of human C-peptide in biological samples This electrochemiluminescence-based assay offers high sensitivity and specificity for diabetes research and clinical applications The kit provides reliable detection of C-peptide a biomarker for endogenous insulin production enabling researchers to assess pancreatic beta-cell function in various sample types including serum and plasma