Learn More
Meso Scale Discovery UPLEX HUM CRP ASY384well
SDP
Catalog No. 5000080658
Encompass
The Meso Scale Discovery U-PLEX Human CRP 384-Well Assay is an electrochemiluminescent immunoassay for quantifying C-reactive protein (CRP) in human serum plasma and cell culture supernatants This assay uses MSD s SULFO-TAG labels and multi-array technology for sensitive CRP measurement with a dynamic range of 16 pg/mL to 10 000 pg/mL It requires only 25 L of sample and offers high reproducibility with coefficients of variation 10% The U-PLEX Human CRP Assay is a simple rapid and cost-effective method for assessing systemic inflammation by measuring CRP levels
Catalog No. 50-000-80658 Supplier Meso Scale Discovery Supplier No. K251L9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human CRP 384-Well Assay is an electrochemiluminescent immunoassay for quantifying C-reactive protein (CRP) in human serum plasma and cell culture supernatants This assay uses MSD s SULFO-TAG labels and multi-array technology for sensitive CRP measurement with a dynamic range of 16 pg/mL to 10 000 pg/mL It requires only 25 L of sample and offers high reproducibility with coefficients of variation 10% The U-PLEX Human CRP Assay is a simple rapid and cost-effective method for assessing systemic inflammation by measuring CRP levels