Learn More
Meso Scale Discovery UPLEX HUM CTACK ASY384well
SDP
Catalog No. 5000080982
Encompass
The Meso Scale Discovery CTACK Kit is an electrochemiluminescent immunoassay for the quantitative measurement of CTACK in human serum plasma and cell culture supernatants It utilizes MSD s patented SULFO-TAG labels and multi-array plate technology to provide a rapid ultra-sensitive and cost-effective method for measuring CTACK with a detection range of 0 06-2500 pg/mL and an assay time of 3 5 hours The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and assay diluent This assay allows high-throughput quantification of CTACK levels for research applications
Catalog No. 50-000-80982 Supplier Meso Scale Discovery Supplier No. K251VDK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery CTACK Kit is an electrochemiluminescent immunoassay for the quantitative measurement of CTACK in human serum plasma and cell culture supernatants It utilizes MSD s patented SULFO-TAG labels and multi-array plate technology to provide a rapid ultra-sensitive and cost-effective method for measuring CTACK with a detection range of 0 06-2500 pg/mL and an assay time of 3 5 hours The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and assay diluent This assay allows high-throughput quantification of CTACK levels for research applications