Learn More
Meso Scale Discovery UPLEX HUM CTLA4 ASY 25 PL
SDP
Catalog No. 5000079220
Encompass
The Meso Scale Discovery U-PLEX Human CTLA-4 Assay Kit is a multiplex immunoassay for measuring CTLA-4 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and precision The assay requires 50 L sample volume and takes 3 5 hours It includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG Read Buffer Wash Buffer Blocker A and calibrators It tests up to 80 samples in duplicate with a detection limit of 0 056 ng/mL
Catalog No. 50-000-79220 Supplier Meso Scale Discovery Supplier No. K151R7K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human CTLA-4 Assay Kit is a multiplex immunoassay for measuring CTLA-4 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and precision The assay requires 50 L sample volume and takes 3 5 hours It includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG Read Buffer Wash Buffer Blocker A and calibrators It tests up to 80 samples in duplicate with a detection limit of 0 056 ng/mL