Learn More
Meso Scale Discovery UPLEX HUM CTLA4 ASY384well
SDP
Catalog No. 5000079570
Encompass
The Meso Scale Discovery U-PLEX Human CTLA-4 384-Well Assay Kit is a multiplex immunoassay that measures CTLA-4 levels in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay with high sensitivity broad dynamic range and low sample consumption The assay requires 10 L of sample per well and can detect CTLA-4 across a range of 156 pg/mL to 100 000 pg/mL It includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels and calibrator containing recombinant CTLA-4 The assay takes approximately 3 hours to complete
Catalog No. 50-000-79570 Supplier Meso Scale Discovery Supplier No. K251R7K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human CTLA-4 384-Well Assay Kit is a multiplex immunoassay that measures CTLA-4 levels in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay with high sensitivity broad dynamic range and low sample consumption The assay requires 10 L of sample per well and can detect CTLA-4 across a range of 156 pg/mL to 100 000 pg/mL It includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels and calibrator containing recombinant CTLA-4 The assay takes approximately 3 hours to complete