Learn More
Meso Scale Discovery UPLEX HUM CTLA4 ASY384well
SDP
Catalog No. 5000079862
Encompass
The Meso Scale Discovery U-PLEX Human CTLA-4 384-Well Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of CTLA-4 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide a highly sensitive and reproducible assay in a 384-well plate format The kit includes a CTLA-4 calibrator SULFO-TAG labeled anti-CTLA-4 detection antibody and 384-well plates pre-coated with capture antibody The assay has a typical sensitivity of 0 08 ng/mL and an upper limit of quantification of 2500 pg/mL It requires 2 5 hours total assay time
Catalog No. 50-000-79862 Supplier Meso Scale Discovery Supplier No. K251R7K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human CTLA-4 384-Well Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of CTLA-4 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide a highly sensitive and reproducible assay in a 384-well plate format The kit includes a CTLA-4 calibrator SULFO-TAG labeled anti-CTLA-4 detection antibody and 384-well plates pre-coated with capture antibody The assay has a typical sensitivity of 0 08 ng/mL and an upper limit of quantification of 2500 pg/mL It requires 2 5 hours total assay time