Learn More
Meso Scale Discovery UPLEX HUM CTLA4 ASYQP 25 PL
SDP
Catalog No. 5000079219
Encompass
The Meso Scale Discovery U-PLEX Human CTLA-4 Assay Kit is a multiplex immunoassay that quantitatively measures CTLA-4 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluent It can accurately quantify CTLA-4 levels with minimal hands-on time using an MSD instrument
Catalog No. 50-000-79219 Supplier Meso Scale Discovery Supplier No. K151R7K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human CTLA-4 Assay Kit is a multiplex immunoassay that quantitatively measures CTLA-4 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluent It can accurately quantify CTLA-4 levels with minimal hands-on time using an MSD instrument