Learn More
Meso Scale Discovery UPLEX HUM Cystatin C ASY384
SDP
Catalog No. 5000080466
Encompass
The Meso Scale Discovery U-PLEX Human Cystatin C 384-Well Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Cystatin C in human serum plasma and urine It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide a rapid high sensitivity multiplex immunoassay requiring small sample volumes with a broad dynamic range This 384-well assay format allows high-throughput analysis of Cystatin C an important biomarker for kidney function The assay has a lower limit of detection of 17 pg/mL and an upper limit of quantification of 250 000 pg/mL
Catalog No. 50-000-80466 Supplier Meso Scale Discovery Supplier No. K251M9K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Cystatin C 384-Well Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Cystatin C in human serum plasma and urine It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide a rapid high sensitivity multiplex immunoassay requiring small sample volumes with a broad dynamic range This 384-well assay format allows high-throughput analysis of Cystatin C an important biomarker for kidney function The assay has a lower limit of detection of 17 pg/mL and an upper limit of quantification of 250 000 pg/mL