Learn More
Meso Scale Discovery UPLEX HUM Cystatin C ASYQP
SDP
Catalog No. 5000080801
Encompass
The Meso Scale Discovery U-PLEX Human Cystatin C Assay Kit quantifies Cystatin C in human serum plasma urine and cell culture supernatants using electrochemiluminescent detection It features a plate-based ELISA format with high sensitivity and a broad dynamic range The kit includes a 96-well plate capture and detection antibodies Read Buffer Sample and Calibrator Diluents and a lyophilized U-PLEX Calibrator Sensitivity is 0 3 ng/mL with an upper limit of 2500 ng/mL It performs up to 90 tests in duplicate in 3 5 hours offering a simple rapid and sensitive measurement method
Catalog No. 50-000-80801 Supplier Meso Scale Discovery Supplier No. K151M9K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Cystatin C Assay Kit quantifies Cystatin C in human serum plasma urine and cell culture supernatants using electrochemiluminescent detection It features a plate-based ELISA format with high sensitivity and a broad dynamic range The kit includes a 96-well plate capture and detection antibodies Read Buffer Sample and Calibrator Diluents and a lyophilized U-PLEX Calibrator Sensitivity is 0 3 ng/mL with an upper limit of 2500 ng/mL It performs up to 90 tests in duplicate in 3 5 hours offering a simple rapid and sensitive measurement method