Learn More
Meso Scale Discovery UPLEX HUM DPPIV ASY384well
SDP
Catalog No. 5000080750
Encompass
The Meso Scale Discovery U-PLEX Human DPP4 384-Well Assay Kit is a multiplex immunoassay that measures DPP4 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This 384-well plate assay allows for high throughput analysis of up to 80 samples in duplicate The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent label calibrator diluent wash buffer read buffer and a 384-well high bind microplate
Catalog No. 50-000-80750 Supplier Meso Scale Discovery Supplier No. K251D9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human DPP4 384-Well Assay Kit is a multiplex immunoassay that measures DPP4 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This 384-well plate assay allows for high throughput analysis of up to 80 samples in duplicate The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent label calibrator diluent wash buffer read buffer and a 384-well high bind microplate