Learn More
Meso Scale Discovery UPLEX HUM DPPIV ASYQP 25 PL
SDP
Catalog No. 5000078743
Encompass
The Meso Scale Discovery U-PLEX Human DPP4 Assay is a multiplex immunoassay panel that measures Dipeptidyl Peptidase 4 (DPP4) in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The U-PLEX assay measures DPP4 levels quantitatively with precision accuracy and reproducibility This immunoassay panel is for research use only
Catalog No. 50-000-78743 Supplier Meso Scale Discovery Supplier No. K151D9K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human DPP4 Assay is a multiplex immunoassay panel that measures Dipeptidyl Peptidase 4 (DPP4) in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The U-PLEX assay measures DPP4 levels quantitatively with precision accuracy and reproducibility This immunoassay panel is for research use only