Learn More
Meso Scale Discovery UPLEX HUM ENA78 ASY 25 PL
SDP
Catalog No. 5000082361
Encompass
The Meso Scale Discovery U-PLEX Human ENA 7-Plex Assay Kit (K151VEK) is an electrochemiluminescent immunoassay for the quantitative measurement of autoantibodies to 7 extractable nuclear antigens (ENA) RNP Sm SS-A SS-B Scl-70 Jo-1 and CENP-B in human serum plasma and other biological fluids This multiplex assay allows for the simultaneous detection of autoantibodies associated with several connective tissue diseases from a single small sample volume of 25 L The kit includes a 96-well plate coated with antigens detection antibody read buffer wash buffer calibrator and controls The assay has a testing time of approximately 3 5 hours and a typical sensitivity of 1 U/mL
Catalog No. 50-000-82361 Supplier Meso Scale Discovery Supplier No. K151VEK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human ENA 7-Plex Assay Kit (K151VEK) is an electrochemiluminescent immunoassay for the quantitative measurement of autoantibodies to 7 extractable nuclear antigens (ENA) RNP Sm SS-A SS-B Scl-70 Jo-1 and CENP-B in human serum plasma and other biological fluids This multiplex assay allows for the simultaneous detection of autoantibodies associated with several connective tissue diseases from a single small sample volume of 25 L The kit includes a 96-well plate coated with antigens detection antibody read buffer wash buffer calibrator and controls The assay has a testing time of approximately 3 5 hours and a typical sensitivity of 1 U/mL