Learn More
Meso Scale Discovery UPLEX HUM ENA78 ASY384well
SDP
Catalog No. 5000080176
Encompass
The Meso Scale Discovery U-PLEX Human ENA 7-Plex Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of 7 autoantibodies associated with systemic autoimmune rheumatic diseases It measures anti-dsDNA anti-Sm anti-RNP anti-Ro60 anti-Ro52 anti-SSA and anti-SSB antibodies in human serum plasma and other biological fluids The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements This multiplex kit allows efficient autoantibody profiling relevant to SLE and other connective tissue diseases
Catalog No. 50-000-80176 Supplier Meso Scale Discovery Supplier No. K251VEK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human ENA 7-Plex Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of 7 autoantibodies associated with systemic autoimmune rheumatic diseases It measures anti-dsDNA anti-Sm anti-RNP anti-Ro60 anti-Ro52 anti-SSA and anti-SSB antibodies in human serum plasma and other biological fluids The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements This multiplex kit allows efficient autoantibody profiling relevant to SLE and other connective tissue diseases