Learn More
Meso Scale Discovery UPLEX HUM ENA78 ASY384well
SDP
Catalog No. 5000081084
Encompass
The Meso Scale Discovery U-PLEX Human ENA 7-Plex Assay is a 384-well electrochemiluminescent immunoassay that measures 7 autoantibodies linked to systemic autoimmune rheumatic diseases in human serum plasma and cell culture supernatant samples It detects IgG autoantibodies to Ro52 Ro60 La Sm RNP Scl-70 and Jo-1 antigens using MSD GOLD technology with carbon electrodes The assay takes about 3 5 hours and shows no cross-reactivity between antigens offering a sensitive reproducible method to measure autoantibody profiles in systemic autoimmune diseases
Catalog No. 50-000-81084 Supplier Meso Scale Discovery Supplier No. K251VEK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human ENA 7-Plex Assay is a 384-well electrochemiluminescent immunoassay that measures 7 autoantibodies linked to systemic autoimmune rheumatic diseases in human serum plasma and cell culture supernatant samples It detects IgG autoantibodies to Ro52 Ro60 La Sm RNP Scl-70 and Jo-1 antigens using MSD GOLD technology with carbon electrodes The assay takes about 3 5 hours and shows no cross-reactivity between antigens offering a sensitive reproducible method to measure autoantibody profiles in systemic autoimmune diseases