Learn More
Meso Scale Discovery UPLEX HUM Eotaxin2 ASY 25 PL
SDP
Catalog No. 5000079631
Encompass
The Meso Scale Discovery U-PLEX Human Eotaxin-2 Assay Kit is designed for the quantitative measurement of human Eotaxin-2 in serum plasma and cell culture supernatants using an electrochemiluminescent detection method The assay uses MSD GOLD Streptavidin-coated plates and a U-PLEX linker to capture and detect bound analyte The kit includes a 96-well plate calibrator diluent wash buffer read buffer 10X eotaxin-2 detection antibody 10X SULFO-TAG streptavidin and a vial of lyophilized calibrator (reconstitute before use) The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL
Catalog No. 50-000-79631 Supplier Meso Scale Discovery Supplier No. K151XQK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Eotaxin-2 Assay Kit is designed for the quantitative measurement of human Eotaxin-2 in serum plasma and cell culture supernatants using an electrochemiluminescent detection method The assay uses MSD GOLD Streptavidin-coated plates and a U-PLEX linker to capture and detect bound analyte The kit includes a 96-well plate calibrator diluent wash buffer read buffer 10X eotaxin-2 detection antibody 10X SULFO-TAG streptavidin and a vial of lyophilized calibrator (reconstitute before use) The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL