Learn More
Meso Scale Discovery UPLEX HUM Eotaxin2 ASY384well
SDP
Catalog No. 5000081037
Encompass
The Meso Scale Discovery U-PLEX Human Eotaxin-2 384-well Assay is a multiplex immunoassay kit that enables the simultaneous quantitative measurement of human Eotaxin-2 in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption This 384-well plate assay requires just 50 L of sample per well and can perform up to 40 tests across two plates The kit includes a plate coated with capture antibodies detection antibodies Read Buffer Sample Diluent and calibrators and controls to generate a standard curve and validate assay performance
Catalog No. 50-000-81037 Supplier Meso Scale Discovery Supplier No. K251XQK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Eotaxin-2 384-well Assay is a multiplex immunoassay kit that enables the simultaneous quantitative measurement of human Eotaxin-2 in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption This 384-well plate assay requires just 50 L of sample per well and can perform up to 40 tests across two plates The kit includes a plate coated with capture antibodies detection antibodies Read Buffer Sample Diluent and calibrators and controls to generate a standard curve and validate assay performance