Learn More
Meso Scale Discovery UPLEX HUM Eotaxin2 ASY384well
SDP
Catalog No. 5000081132
Encompass
The Meso Scale Discovery U-PLEX Human Eotaxin-2 384-well Assay is a multiplex immunoassay kit that enables simultaneous quantification of human Eotaxin-2 in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This 384-well plate assay allows high-throughput analysis of Eotaxin-2 a chemokine involved in eosinophil chemotaxis during allergic inflammatory responses With a measuring time of just 3 5 hours it offers rapid and accurate quantification of Eotaxin-2 levels
Catalog No. 50-000-81132 Supplier Meso Scale Discovery Supplier No. K251XQK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Eotaxin-2 384-well Assay is a multiplex immunoassay kit that enables simultaneous quantification of human Eotaxin-2 in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This 384-well plate assay allows high-throughput analysis of Eotaxin-2 a chemokine involved in eosinophil chemotaxis during allergic inflammatory responses With a measuring time of just 3 5 hours it offers rapid and accurate quantification of Eotaxin-2 levels