Learn More
Meso Scale Discovery UPLEX HUM Eotaxin3 ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K151UEK4
Encompass
The Meso Scale Discovery U-PLEX Human Eotaxin-3 Assay Kit is designed for the quantitative measurement of human Eotaxin-3 in serum plasma and cell culture supernatants using an electrochemiluminescent detection method The assay uses MSD s proprietary QuickPlex SQ 120 instrument and provides high sensitivity and a broad dynamic range This multiplex assay allows users to measure Eotaxin-3 along with other biomarkers in the same sample The kit includes a plate coated with capture antibodies detection antibodies calibrators and controls needed to perform the assay
Catalog No. 50-000-79037
Add to Cart
Request a Quote