Learn More
Meso Scale Discovery UPLEX HUM Eotaxin3 ASYQP
SDP
Catalog No. 5000079714
Encompass
The MSD U-PLEX Human Eotaxin-3 Singleplex Assay is an electrochemiluminescence-based immunoassay designed for quantitative measurement of eotaxin-3 (CCL26) protein levels in human biological samples This singleplex format allows focused analysis of this specific chemokine involved in eosinophil recruitment and allergic inflammation The U-PLEX platform offers high sensitivity wide dynamic range and minimal sample volume requirements making it suitable for research applications in immunology allergy studies and inflammatory disease research
Catalog No. 50-000-79714 Supplier Meso Scale Discovery Supplier No. K151UEK24
Add to Cart
Add to Cart
Request a Quote
The MSD U-PLEX Human Eotaxin-3 Singleplex Assay is an electrochemiluminescence-based immunoassay designed for quantitative measurement of eotaxin-3 (CCL26) protein levels in human biological samples This singleplex format allows focused analysis of this specific chemokine involved in eosinophil recruitment and allergic inflammation The U-PLEX platform offers high sensitivity wide dynamic range and minimal sample volume requirements making it suitable for research applications in immunology allergy studies and inflammatory disease research