Learn More
Meso Scale Discovery UPLEX HUM ESelectin ASY
SDP
Catalog No. 5000081585
Encompass
The Meso Scale Discovery U-PLEX Human E-Selectin Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of E-Selectin in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can perform up to 80 tests across two plates The lower limit of detection is 0 05 ng/mL This product can help researchers investigate E-Selectin s role in inflammation endothelial cell activation and leukocyte trafficking
Catalog No. 50-000-81585 Supplier Meso Scale Discovery Supplier No. K151AGVK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human E-Selectin Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of E-Selectin in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can perform up to 80 tests across two plates The lower limit of detection is 0 05 ng/mL This product can help researchers investigate E-Selectin s role in inflammation endothelial cell activation and leukocyte trafficking