Learn More
Meso Scale Discovery UPLEX HUM ESelectin ASY384
SDP
Catalog No. 5000080986
Encompass
The Meso Scale Discovery U-PLEX Human E-Selectin Assay is a 384-well multiplex immunoassay kit for the quantitative measurement of E-Selectin in human serum plasma and cell culture supernatants The assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to a 7-log dynamic range and low sample volume requirements This assay allows for the simultaneous analysis of E-Selectin and other biomarkers It includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance
Catalog No. 50-000-80986 Supplier Meso Scale Discovery Supplier No. K251AGVK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human E-Selectin Assay is a 384-well multiplex immunoassay kit for the quantitative measurement of E-Selectin in human serum plasma and cell culture supernatants The assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to a 7-log dynamic range and low sample volume requirements This assay allows for the simultaneous analysis of E-Selectin and other biomarkers It includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance