Learn More
Meso Scale Discovery UPLEX HUM ESelectin ASYQP
SDP
Catalog No. 5000080470
Encompass
The Meso Scale Discovery U-PLEX Human E-Selectin Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of E-Selectin in human serum plasma and cell culture supernatants It utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer and plate seals It can accurately quantify E-Selectin levels between 0 05-500 ng/mL with intra-assay precision of 10% CV
Catalog No. 50-000-80470 Supplier Meso Scale Discovery Supplier No. K151AGVK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human E-Selectin Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of E-Selectin in human serum plasma and cell culture supernatants It utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer and plate seals It can accurately quantify E-Selectin levels between 0 05-500 ng/mL with intra-assay precision of 10% CV