Learn More
Meso Scale Discovery UPLEX HUM Factor VII ASY
SDP
Catalog No. 5000082331
Encompass
The Meso Scale Discovery U-PLEX Human Factor VII Assay Kit is an electrochemiluminescent immunoassay for measuring Factor VII in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive detection The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and diluent Sensitivity is 0 05 ng/mL with a quantification limit of 1500 ng/mL Results are ready in 3 5 hours using an MSD instrument This assay supports high-throughput analysis in research and drug development
Catalog No. 50-000-82331 Supplier Meso Scale Discovery Supplier No. K151N9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Factor VII Assay Kit is an electrochemiluminescent immunoassay for measuring Factor VII in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive detection The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and diluent Sensitivity is 0 05 ng/mL with a quantification limit of 1500 ng/mL Results are ready in 3 5 hours using an MSD instrument This assay supports high-throughput analysis in research and drug development