Learn More
Meso Scale Discovery UPLEX HUM Factor VII ASY384
SDP
Catalog No. 5000081044
Encompass
The Meso Scale Discovery U-PLEX Human Factor VII 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of Factor VII in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide a sensitive and reproducible assay with a broad dynamic range This multiplexed assay allows high-throughput analysis of Factor VII levels with low sample volume requirements The kit includes a 384-well plate coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay
Catalog No. 50-000-81044 Supplier Meso Scale Discovery Supplier No. K251N9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Factor VII 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of Factor VII in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide a sensitive and reproducible assay with a broad dynamic range This multiplexed assay allows high-throughput analysis of Factor VII levels with low sample volume requirements The kit includes a 384-well plate coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay