Learn More
Meso Scale Discovery UPLEX HUM FGF basic ASY
SDP
Supplier:  Meso Scale Discovery K151X7K4
Encompass
The Meso Scale Discovery U-PLEX Human FGF Basic Assay is a multiplex immunoassay kit for quantifying human FGF basic in various biological matrices It uses electrochemiluminescence detection offering high sensitivity a dynamic range of up to 5 logs and low sample consumption Requiring 50 L of sample it is compatible with Meso Scale Discovery instruments like the MESO QuickPlex SQ 120 series The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent tags and calibrators and controls for standard curve generation and assay validation
Catalog No. 50-000-82402
Add to Cart
Request a Quote