Learn More
Meso Scale Discovery UPLEX HUM FGF basic ASY384
SDP
Catalog No. 5000081124
Encompass
The Meso Scale Discovery U-PLEX Human FGF Basic 384-Well Assay is a multiplex immunoassay for measuring FGF basic in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a dynamic range of up to 5 logs and low sample consumption The assay measures FGF basic levels from 0 06 to 2500 pg/mL with just 10 L of sample per well Provided in a 384-well plate it includes analyte-specific antibodies on carbon electrodes and takes about 3 hours to complete enabling sensitive reproducible high-throughput quantification from small samples
Catalog No. 50-000-81124 Supplier Meso Scale Discovery Supplier No. K251X7K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human FGF Basic 384-Well Assay is a multiplex immunoassay for measuring FGF basic in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a dynamic range of up to 5 logs and low sample consumption The assay measures FGF basic levels from 0 06 to 2500 pg/mL with just 10 L of sample per well Provided in a 384-well plate it includes analyte-specific antibodies on carbon electrodes and takes about 3 hours to complete enabling sensitive reproducible high-throughput quantification from small samples